This product is now discontinued, View Related Products
IKA Magic Lab Mixing Heads
Interchangeable mixing heads improve versatility.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Jar Capacity (L) Speed (rpm)
Availability Pricing
IKA R078352 Three-stage high shear dispersing mixing head for 04300-01 Magic Lab EW-04201-21 Mfr # U078352
COMPARE
80 3,000 to 26,000
Call For Price
IKA R076662 Colloid mill mixing head for 04300-01 Magic Lab (sold separately) EW-04201-23 Mfr # U076662
COMPARE
700 3,000 to 26,000
Call For Price
IKA R079664 Cone mill mixing head for 04300-01 Magic Lab (sold separately) EW-04201-25 Mfr # U076664
COMPARE
700 3,000 to 26,000
Call For Price
IKA R075333 Powder-liquid dispersing mixing head for 04300-01 Magic Lab (sold separately) EW-04201-27 Mfr # R075333
COMPARE
800 3,000 to 16,000
Call For Price
IKA R075262 Suction-feed powder-liquid dispersing mixing head for 04300-01 Magic Lab (sold separately) EW-04201-29 Mfr # R075262
COMPARE
60 3,000 to 26,000
Call For Price
IKA R079665 Micro-plant mixing head for 04300-01 Magic Lab; and 04201-21, -23, -25 Mixing Heads EW-04201-31 Mfr # U079665
COMPARE
Based on mixing head used
Call For Price
IKA R079766 Batch dispersing mixing head for 04300-01 Magic Lab (sold separately) EW-04201-33 Mfr # R079766
COMPARE
1.5 10,000 to 24,000
Call For Price
Showing 1- 7 of 7