IKA T 65 digital ULTRA-TURRAX Batch Process Homogenizer
High-performance dispersing designed for typical pilot plant volumes
  • Process volumes from 2 to 50 liters
  • Plug-in connectors facilitate exchange of dispersing elements
With a digital speed display, the T 65 D homogenizer offers reproducible operations even with changes in viscosities since the speed remains constant. The stainless steel dispersing elements can be used under vacuum and pressure and come in three configurations to meet a wide range of applications (see our web site for additional configurations). The wide speed range, from 1000-9500 rpm, allows for work at high circumferential speeds, even with small rotor diameters. Safety features include an electronic overload protection and error code display.
A complete system is composed of the homogenizer, rotor-stator, and stand.
$11,757.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Sample Size (mL) Max Speed (rpm) Min Sample Size (mL)
Availability Pricing
IKA T 65 D Ultra-Turrax Batch Process Homogenizer; 230-400 VAC, 50/60 Hz, 3 Ph EW-04737-33 Mfr # 4234501
COMPARE
50000 9500 2000
$11,757.00USD / Each