Imada Durometer EX Hardness Tester
Won’t destroy your sample
  • Heavy-duty units feature well-made design with high repeatability
  • Measurement scale is 100 points with an accuracy of ±1 point
Durometers feature portable design, ease of operation, and non-destructive measurement. They are heavy-duty and well-made with high repeatability. The measurement of hardness is an essential factor for determining the characteristics of various rubber, elastomers and plastic products. One of the most important steps in using a hardness tester is to select the optimum type from the various types available based upon different material hardnesses. The measurement scale is 100 points with an accuracy of ± 1 point.
Hardness Tester Selection
  • Model D: Hard rubber and plastics
  • Model A: Normal rubber and softer plastics
  • Asker C: Soft rubber and sponge
$530.50 - $546.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Type
Availability Pricing
Imada EX-2 Durometer Hardness Tester for Normal Rubber, Shore A EW-59792-35 Mfr # EX-2
COMPARE
Shore A
$546.50USD / Each
Imada EX-D Durometer for Hard Rubber and Plastics EW-59792-37 Mfr # EX-D
COMPARE
Shore D
$530.50USD / Each