Benchmark Scientific Incu-Shaker™ Mini Incubating Shaker
Instantly change flask clamps with unique magnetic platform and accessories
  • Clamps use a powerful magnetic base to securely attach to the shaking platform without screws, bolts, or tools
  • Small footprint saves valuable benchtop space
  • Microprocessor controlled shaking speed, temperature, and run time ensures accuracy
  • Easy-to-read full color, touchscreen display
This space-saving incubating shaker offers exceptional temperature uniformity and accuracy. Proprietary heat distribution technology ensures temperatures are precisely maintained throughout the chamber to within 0.25%, even in the corners. Digitally regulated temperature with precisely controlled orbital mixing is optimized for culturing a wide variety of cell types.
This shaker lets you customize the size and types of vessels being used by simply exchanging clamp sizes—purchase clamps separately to suit your application needs. Speed, temperature, and time are controlled with a touchscreen display. Overtemperature and overspeed controls ensure safety, long life, and sample integrity.
A good-sized work area (11-1/2" x 9-1/2"/29.2 x 24.1 cm) in a compact footprint is perfect for the benchtop. Brushless induction drive provides durability and longevity while noise and vibration are reduced thanks to a motor isolation system.
$3,774.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Benchmark Scientific H-1001-M with Magic Clamp Platform, 115 V EW-51705-01 Mfr # H1001-M-CP
COMPARE
115
$3,774.00USD / Each
Benchmark Scientific H-1001-M-E with Magic Clamp Platform, 230 V EW-51705-03 Mfr # H1001-M-CP-E
COMPARE
230
$3,774.00USD / Each