Great Plains Industries Industrial Gear Flowmeters
High-accuracy measurement of low and high viscosity liquids
  • No requirements for straight pipe runs
  • Only two moving parts in meter
  • High repeatability and direct volumetric reading
With flow rates from 0.5 GPH to 40 GPM, viscosities up to 1000 cps and three housing materials, these meters handle most metering applications. This positive displacement oval gear meter does not require calibration and is accurate to ±1% of reading. Loop-powered 4 to 20 mA output uses three buttons for simple and easy setting of 4 and 20 mA signals.
Aluminum material is recommended for petroleum products.
Choose stainless steel and PPS housings for products ranging from water to petroleum products to aggressive acids and caustics. Meters are designed for use with liquids such as solvents, fuel, catalysts, oils, grease, and even adhesives — making them perfect for the mining, petroleum, paint, and chemical industries.
$765.00 - $1,647.50USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Pipe Size Wetted Materials Description
Availability Pricing
Great Plains Industries OM00 Gear Flowmeter, SS, 1/4" Process Connections with Reed/Hall Output EW-32930-01 Mfr # OM006S513-222
COMPARE
1/4" SS Gear Flowmeter, SS, 1/4" Process Connections with Reed/Hall Output
$914.00USD / Each
Great Plains Industries OM00 Gear Flowmeter, SS, 3/8" Process Connections with Reed/Hall Output EW-32930-11 Mfr # OM008S513-222
COMPARE
3/8" SS Gear Flowmeter, SS, 3/8" Process Connections with Reed/Hall Output
$967.00USD / Each
Great Plains Industries OM01 Gear Flowmeter, Al, 1/2" Process Connections with Reed/Hall Output EW-32930-19 Mfr # OM015A001-222
COMPARE
1/2" Aluminum Gear Flowmeter, Al, 1/2" Process Connections with Reed/Hall Output
$765.00USD / Each
Great Plains Industries OM01 Gear Flowmeter, SS, 1/2" Process Connections with Reed/Hall Output EW-32930-27 Mfr # OM015S513-222
COMPARE
1/2" SS Gear Flowmeter, SS, 1/2" Process Connections with Reed/Hall Output
$1,647.50USD / Each
Great Plains Industries OM02 Gear Flowmeter, PPS, 1" Process Connections with Reed/Hall Output EW-32930-35 Mfr # OM025P003-822
COMPARE
1" PPS Gear Flowmeter, PPS, 1" Process Connections with Reed/Hall Output
$863.00USD / Each