This product is now discontinued, View Related Products
Infrared PVDF Flow Sensors
Place in any orientation without flow straighteners
These infrared sensors feature a unique turbine design that prevents air or gas bubbles from being trapped in the measuring chamber leading to improved accuracy and repeatability. The design also utilizes a fluid bearing and operates virtually friction free minimizing component wear. The rotor assembly is removable for cleaning or replacement of the flow tube. For protection from particulate, models 32250-12 through -52 include a 100 ìm filter screen; additional screening may be required for -02 and -12 models, see specifications. Note: these sensors cannot be used with opaque fluids.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Min Flow Rate (GPM) Process Connection
Availability Pricing
JLC PVDF Infrared Flow Sensor; 0.0. to 0.53 GPM, 1/4" barb connection EW-32250-02 Mfr # IR-OPFLOW-100.11H
COMPARE
0.53 0.03 1/4" Hose Barb
Call For Price
JLC PVDF Infrared Flow Sensor; 0.08 to 2.38 GPM, 3/8" barb connection EW-32250-12 Mfr # IR-OPFLOW-100.21H
COMPARE
2.38 0.08 3/8" Hose Barb
Call For Price
JLC PVDF Infrared Flow Sensor; 0.13 to 3.96 GPM, 7/16" barb connection EW-32250-22 Mfr # IR-OPFLOW-100.31H
COMPARE
3.96 0.13 7/16" Hose Barb
Call For Price
JLC PVDF Infrared Flow Sensor; 0.26 to 7.93 GPM, 5/8" barb connection EW-32250-32 Mfr # IR-OPFLOW-100.41H
COMPARE
7.93 0.26 5/8" Hose Barb
Call For Price
JLC PVDF Infrared Flow Sensor; 0.66 to 19.8 GPM, 3/4" barb connection EW-32250-42 Mfr # IR-OPFLOW-100.51H
COMPARE
19.8 0.66 3/4" Hose Barb
Call For Price
JLC PVDF Infrared Flow Sensor; 1.06 to 32 GPM, 7/8" barb connection EW-32250-52 Mfr # IR-OPFLOW-100.61H
COMPARE
32 1.06 7/8" Hose Barb