This product is now discontinued, View Related Products
Instek Precision LCR Meter
Single-layer menu makes set up and operation quick and easy
  • Wide test frequency 20 Hz to 1, 5, and 10 MHz
  • Measures 11 different parameters with 0.1% basic accuracy
  • 6-digit measurement resolution
  • Large LCD display with intuitive user interface
  • Full measuring functions with DUT V/I monitor
  • Pass/fail function with judgment alarm
  • DC resistance measurement
  • Multi-step mode
  • Graph mode for visual characterization
  • RS-232C and GPIB interface standard
These LCR meters provide accuracy, versatility, and high resolution, for a wide range of component measurements, including DC resistance measurement and voltage/current monitoring. Use the multi-step function for on-screen programming of customized measurement sequences with pass/fail indication. Each program includes 30 test steps and each test step can be set with selected parameters and test limits. These meters automate tedious manual work routines with the press of a button.
In graph mode, select the parameter, set the start/stop voltage or the start/stop frequency of the sweep, and meters run through the sequence of measurements displaying the component characteristics in graphical format. The standard GPIB and RS-232C interfaces provide easy instrument control and downloadable results to your PC.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Test Frequency
Availability Pricing
GW Instek LCR-8101G 20 Hz to 1 MHz Precision LCR Meter EW-20036-57 Mfr # LCR-8101G
COMPARE
20 Hz to 1 MHz, 20 Hz to 5 MHz, 20 Hz to 10 MHz
Call For Price
GW Instek LCR-8105G 20 Hz to 5 MHz Precision LCR Meter EW-20036-58 Mfr # LCR-8105G
COMPARE
20 Hz to 1 MHz, 20 Hz to 5 MHz, 20 Hz to 10 MHz
Call For Price
GW Instek LCR-8110G 20 Hz to 10 MHz Precision LCR Meter EW-20036-59 Mfr # LCR-8110G
COMPARE
20 Hz to 1 MHz, 20 Hz to 5 MHz, 20 Hz to 10 MHz