Iodine Solution 23255

$67.00 - $741.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Fisher Chemical SI1064 Iodine Solution (Wijs) (4 liter) EW-88084-75 Mfr # SI1064
COMPARE
7553-56-2 and 64-19-7 and 7782-50-5 4000 Iodine Solution (Wijs) (4 liter)
$741.00USD / Each
Fisher Chemical SI106500 Iodine Solution (Wijs) (500mL) CAS 7553-56-2 EW-88084-76 Mfr # SI106500
COMPARE
7553-56-2 and 64-19-7 and 7782-50-5 500 Iodine Solution (Wijs) (500mL) CAS 7553-56-2
$149.00USD / Each
COMPARE
7553-56-2, 7681-11-0, 7732-18-5 946 Iodine (Iodine-Iodide), 0.100 Normal (N/10) (32 oz)
$67.00USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat