Iridium Flow Controllers
Incorporate into your pump system for complete control
  • Color touchscreen provides control and feedback
  • Very durable, designed for lab to process use
  • Configurable for your changing flow control requirements
The Iridium is a platform for configurable flow control. Control your pump digitally or through voltage for precise flow during pumping and batch flow. Its modular design allows for a single device to adapt to a variety of flow conditions and applications. Standard flow controller features a PTFE microturbine flow sensing module and a PTFE proportional diaphragm flow control module valve to regulate flow rate regardless of inlet pressure fluctuations. This configuration is set up to control liquid flow ranges as low as 7-50 mL/minute and as high as 1.0-10.0L/minute.
A flow control setpoint is provided by a user via the integrated 3.2"H touchscreen, or remotely via USB, RS485, or analog signal. The controller then adjusts the integrated valve as needed to maintain the desired flow rate. The user can program setpoints, alarms, display current status, and adjust configuration while the unit continues to control flow rate and provide output. Where required, the touchscreen can become a digital keypad for simple numeric value entry. For repeatable liquid flow measurement, the flow module uses a miniature PTFE microturbine wheel suspended on a sapphire shaft. An infrared beam detects the rotation speed. This technology is compatible with virtually any low-viscosity fluid and provides fast respnse and long-term reliablilty
RS485 serial interface and USB interfaces are standard, but the Iridium is modular and can be built in many configurations. This supports field-replacement of internal modules as needed, allowing the users to swap flow ranges, valve types, I/O modules, and more as application conditions change. These swaps take only minutes and provide virtually limitless configurations. For more information, contact an Application Specialist.
$5,195.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
McMillan Flow Touchscreen Flow Controller, Viton Seals, 15-100 mL/Minute, 1/8" FNPT Connections, Analog I/O, Alarm EW-32720-00 Mfr # 6010-01-C1-3-N2-EAA-EBA
COMPARE
Touchscreen Flow Controller, Viton Seals, 15-100 mL/Minute, 1/8" FNPT Connections, Analog I/O, Alarm
$5,195.00USD / Each
McMillan Flow Touchscreen Flow Controller, Viton Seals, 50-500 mL/Minute, 1/8" FNPT Connections, Analog I/O, Alarm EW-32720-02 Mfr # 6010-01-C1-3-N2-EAA-EBA
COMPARE
Touchscreen Flow Controller, Viton Seals, 50-500 mL/Minute, 1/8" FNPT Connections, Analog I/O, Alarm
$5,195.00USD / Each
McMillan Flow Touchscreen Flow Controller, Viton Seals, 100-1,000 mL/Minute, 1/8" FNPT Connections, Analog I/O, Alarm EW-32720-04 Mfr # 6010-01-C1-3-N2-EAA-EBA
COMPARE
Touchscreen Flow Controller, Viton Seals, 100-1,000 mL/Minute, 1/8" FNPT Connections, Analog I/O, Alarm
$5,195.00USD / Each
McMillan Flow Touchscreen Flow Controller, Viton Seals, 500-5,000 mL/Minute, 1/4" FNPT Connections, Analog I/O, Alarm EW-32720-06 Mfr # 6010-01-C1-3-N4-EAA-EBA
COMPARE
Touchscreen Flow Controller, Viton Seals, 500-5,000 mL/Minute, 1/4" FNPT Connections, Analog I/O, Alarm
$5,195.00USD / Each