This product is now discontinued, View Related Products
Ismatec® Analog Variable-Speed Multichannel Peristaltic Pumps
Easily adjust speed of compact multichannel pump
  • Start/stop pump or change flow direction via contact closure (DB15 female)
  • Monitor speed through frequency output
These space-saving pumps provide up to four pumping channels while using very little benchtop space. Self-lubricating, Ertalyte® rollers provide consistent flow with reduced tubing wear. Use Click’n’go™ minicartridges for consistent results. Reverse flow direction with the flip of a switch. Select two- or four-channel pumps; twelve roller pumps provide smooth, even flow.
Speed is adjustable in 1% steps (0 to 99%). Control speed remotely with an analog input signal (0 to 5 VDC, 0 to 10 VDC, 0 to 20 mA, or 4 to 20 mA).
Choose optional minicartridges with adjustable occlusion to manually vary pressure on each channel. Pumps are designed to be used with 3-stop tubing. (Scroll down to see all accessory choices.)
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Number of Channels Power (VAC)
Availability Pricing
COMPARE
68 2 115
Call For Price
COMPARE
68 2 230
Call For Price
COMPARE
57 2 115
Call For Price
COMPARE
57 2 230
Call For Price
COMPARE
35 4 115
Call For Price
COMPARE
35 4 230
Call For Price
COMPARE
38 2 115
Call For Price
COMPARE
38 2 230
Call For Price
COMPARE
43 4 115
Call For Price
COMPARE
43 4 230
Call For Price
COMPARE
24 4 115
Call For Price
COMPARE
24 4 230
Call For Price
Showing 1- 12 of 12