Ismatec® BVP-Z Series Analog Gear Pump Drive
Three-digit, push-button potentiometer allows you set 1000:1 turndown ratio
  • Four analog signal ranges and on/off control allow peripheral instruments to adjust speed and actuate drive (remote cable required)
  • Press the MAX button for extra rpm and quick pump priming
  • Electronic feedback system compensates for changes in load to maintain ±1% speed accuracy
Analog input (0 to 5 V, 0 to 10 V, 0 to 20 mA, or 4 to 20 mA) allows for remote speed control. Or, use the unique, three-digit potentometer to set motor speed.
$2,754.00 - $2,946.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Duty Cycle Continuous Max Temperature (° F) Power (VAC)
Availability Pricing
COMPARE
Yes, 1200 to 5000 rpm 104 115
$2,946.00USD / Each
COMPARE
Yes, 1200 to 5000 rpm 104 230
$2,754.00USD / Each