Ismatec® Compact Integrated Drive for Micropump® Pump Heads
High-performance drives with numerous features
  • Control speed and start/stop remotely via analog signal; RS-232 available on Reglo Digital and MCP-series drives Magnetically driven pump heads provide smooth, accurate, pulseless fluid delivery Don't forget your pump head and tubing for a complete pump system Adapter plate to accept either A or B Mount gear pump heads is included
Compatible with all Micropump® A-mount (with adapter plate 07617-90 - included) or B-mount pump heads. Country specific power cord shipped with drive - chosen based upon ship-to address.
Two main gear pump styles fit the drives (pump heads sold separately):
Cavity style pump head can be used for viscous media and for applications with a certain suction lift, has smooth operation at a low noise, and low inner friction.
Suction shoe pump head features enhanced pump performance at elevated differential pressures and has fewer parts for easy maintenance. These pump heads are suitable for elevated temperature ranges and not suitable for applications requiring a suction lift.
Some gear pump head models have an internal fluid bypass. A fluid bypass valve helps protect against decoupling and damage from high pressure build-up.
$2,082.00 - $5,108.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Motor Enclosure Type Power (VAC) Description
Availability Pricing
COMPARE
PMDC 115 MCP Standard Digital Drive for Micropump Pump Heads; 115 VAC
$3,618.00USD / Each
COMPARE
PMDC 230 MCP Standard Digital Drive for Micropump Pump Heads; 230 VAC
$3,618.00USD / Each
COMPARE
PMDC 115 / 230 MCP Process Digital Drive for Micropump Pump Head; 115 to 230 VAC
$5,108.00USD / Each
COMPARE
PMDC 230 Reglo-Z Analog Drive for Micropump Pump Heads; 230 VAC
$2,082.00USD / Each
COMPARE
PMDC 115 / 230 Reglo-Z Programmable Digital Drive for Micropump Pump Heads; 115 to 230 VAC
$2,448.00USD / Each
COMPARE
PMDC 115 / 230 Reglo-ZS Programmable Digital Modular Drive for Micropump Pump Heads; 115 to 230 VAC
$2,906.00USD / Each