Ismatec® Fixed-Speed Rack-Mount Minicartridge Peristaltic Pumps
Compact size mounting rod—use these pumps almost anywhere
  • A constant-speed, synchronous motor ensures a steady, repeatable flow rate
  • Change tubing sizes to obtain different flow rates
Minicartridge design offers the easiest and most convenient tubing changes of all—even replace tubing while your pump is running! Order extra optional minicartridges with adjustable occlusion to manually vary pressure on each channel. Have extra 3-stop tubing on hand when fast tubing changes are crucial. (Scroll down to see all accessory choices.)
$809.00 - $986.00USD / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate Per Channel (mL/min) Number of Rollers Number of Channels
Availability Pricing
COMPARE
6.2 8 2
$875.00USD / Each
COMPARE
6.2 8 2
$875.00USD / Each
COMPARE
$875.00USD / Each
COMPARE
19 8 2
$875.00USD / Each
COMPARE
19 8 2
$875.00USD / Each
COMPARE
6.2 8 4
$986.00USD / Each
COMPARE
6.2 8 4
$986.00USD / Each
COMPARE
12 8 4
$986.00USD / Each
COMPARE
12 8 4
$986.00USD / Each
COMPARE
7.4 6 4
$933.00USD / Each
COMPARE
7.4 6 4
$933.00USD / Each
COMPARE
15 6 4
$933.00USD / Each
COMPARE
15 6 4
$933.00USD / Each
COMPARE
22 6 2
$809.00USD / Each
COMPARE
22 6 2
$809.00USD / Each
COMPARE
7.4 6 2
$809.00USD / Each
COMPARE
7.4 6 2
$809.00USD / Each
COMPARE
7.4 6 2
$809.00USD / Each
COMPARE
7.4 6 2
$809.00USD / Each
Showing 1- 19 of 19