Ismatec® IP and IP-N Digital Peristaltic Pumps
Smooth, precise flow and fewer tubing changes
  • Flow rate 11 to 44 mL/min per channel - flow rate depends on drive rpm│Planetary drive with eight SS rotors reduces pulsation, increases flow rate accuracy, and lengthens tubing life
  • Four-digit LED display facilitates repeatability
  • Control pump via 6-button keypad or analog and TTL external signals│Automatic occlusion cartridges are easy to use and provide reproducible results
These multichannel pumps feature planetary gears that drive eight stainless steel rollers in unison for smooth rotation and accurate liquid flow. The four-digit LED display shows rpm or percent of maximum rpm when pump is running. Use the MAX button to instantly increase speed for priming or flushing without disturbing your programmed settings.
Combine tubing sizes and formulations to suit your application. There are two types of cassettes offered - Click’n’go™ and Pressure Lever cartridges. Click’n’go™ feature automatic occlusion for simplified use, fewer tubing changes, and reproducible results. Install cartridges in one direction for low pulsation; in opposite direction for higher counter pressure. Pressure Lever Cartridges offer the user the opportunity to manually adjust the back pressure experienced by the tubing / pump combination
The pumps with Click'n;go cassettes / cartridges start with 78011-xx.
The pumps with pressure lever cassettes / cartridges start with 78010-xx.
Order the two-stop tubing size and formulation to suit your application (sold separately). More than 100 options exist — the most popular sizes and formulations can be found under Accessories below!
Enter pump speeds via the 6-button keypad for precise pumping—ideal for continuous pumping.
Analog inputs allow you to remotely control speed; TTL contact closure provides start/stop, rotation direction, and autostart control. Monitor speed via analog output. Dispenser may be attached to DB15 female port.
$4,280.00 - $5,947.00USD / Each
35 variations of this product are available.
See All Product Options Below
35 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Max Flow Rate Per Channel (mL/min) Number of Channels
Availability Pricing
COMPARE
115 44 4
$4,392.00USD / Each
COMPARE
230 44 4
$4,392.00USD / Each
COMPARE
115 44 8
$4,768.00USD / Each
COMPARE
230 11 8
$4,768.00USD / Each
COMPARE
230 44 8
$4,768.00USD / Each
COMPARE
230 44 12
$5,024.00USD / Each
COMPARE
115 44 16
$5,504.00USD / Each
COMPARE
115 11 4
$4,280.00USD / Each
COMPARE
115 44 4
$4,370.00USD / Each
COMPARE
230 11 4
$4,280.00USD / Each
COMPARE
230 44 4
$4,370.00USD / Each
COMPARE
115 44 8
$4,727.00USD / Each
COMPARE
230 11 8
$4,768.00USD / Each
COMPARE
230 44 8
$4,727.00USD / Each
COMPARE
115 11 12
$5,025.00USD / Each
COMPARE
115 44 12
$5,094.00USD / Each
COMPARE
230 11 12
$5,025.00USD / Each
COMPARE
115 44 16
$5,516.00USD / Each
COMPARE
115 11 24
$5,824.00USD / Each
COMPARE
230 44 24
$5,912.00USD / Each
Showing 1 - 20 of 35