Ismatec® Microbore Pump Tubing, FDA-Compliant Viton®
Excellent chemical resistance
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Viton® tubing has excellent chemical resistance, especially to acids and alkalis, making it an excellent choice for demanding applications such as acid and solvent transfer. It also handles temperatures from 8 to 400°>F, making it ideal for high-temperature work.
Viton tubing is opaque black. Our Viton is FDA-compliant (21 CFR 177.2600) and is manufactured according to GMP. It is not recommended for sterilization.
Especially designed to be used with Viton color-coded stopped tubing.
Viton Microbore Tubing for Laboratory Equipment and other Microbore Tubing are available to order online. Click any Microbore Tubing category to view all models.
The overall length of Viton® 97632-series extension tubing is 50 feet (15 m). The legacy Ismatec extension tubing was 10 m long.
$155.00 - $161.00USD / Pkg of 1
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
C/L Masterflex tubing size
Availability Pricing
COMPARE
18
$161.00USD / Roll of 1
COMPARE
22
$160.00USD / Roll of 1
COMPARE
24
$159.00USD / Roll of 1
COMPARE
26
$161.00USD / Pkg of 1
COMPARE
28
$155.00USD / Roll of 1
COMPARE
30
$159.00USD / Pkg of 1
COMPARE
32
$156.00USD / Roll of 1
COMPARE
34
$160.00USD / Pkg of 1
COMPARE
36
$155.00USD / Roll of 1
COMPARE
38
$156.00USD / Pkg of 1
COMPARE
40
$156.00USD / Roll of 1
COMPARE
42
$157.00USD / Pkg of 1
COMPARE
44
$155.00USD / Roll of 1
COMPARE
46
$159.00USD / Pkg of 1
COMPARE
48
$160.00USD / Pkg of 1
Showing 1- 15 of 15