Ismatec® Microbore Pump Tubing, Platinum-Cured Silicone
Smooth surface and low protein binding levels
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Use platinum-cured silicone tubing for high purity applications where outgassing of peroxide product is not allowed.
$243.00USD / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
C/L Masterflex tubing size
Availability Pricing
COMPARE
18
$243.00USD / Pkg of 1
COMPARE
26
$243.00USD / Pkg of 1
COMPARE
30
$243.00USD / Pkg of 1
COMPARE
34
$243.00USD / Pkg of 1
COMPARE
42
$243.00USD / Pkg of 1
COMPARE
48
$243.00USD / Pkg of 1