Ismatec® Minicartridge Pump Heads for Masterflex® L/S® Drives
Combine the accuracy of Ismatec® with the versatility of Masterflex® tubing pump systems
  • Order optional minicartridges with adjustable occlusion to vary pressure each channel
  • Easily remove or insert minicartridges without disturbing other channels
  • Max pressure of 1.5 bar is achieved with pressure-lever cassettes only
  • Click-n-go cassettes offer a max pressure of 1.0 bar.
Choose from six, eight, or twelve roller heads. Less rollers offer greater flow rates, while more rollers offer a virtually pulseless flow. Heads accept up to four or eight Click’n’go™ minicartridges (included). Click’n’go minicartridges feature spring occlusion for optimum tension at all times with no adjustments.
Call our Application Specialists to determine the maximum recommended number of cartridges to use with your drive.
$876.00 - $1,035.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Channels Rollers Description
Availability Pricing
COMPARE
8 6 Minicartridge Multichannel Pump Head for Masterflex® L/S® Drives, 8-Channel, 6-Roller
$876.00USD / Each
COMPARE
8 8 Minicartridge Multichannel Pump Head for Masterflex® L/S® Drives, 8-Channel, 8-Roller
$942.00USD / Each
COMPARE
4 12 Minicartridge Multichannel Pump Head for Masterflex® L/S® Drives, 4-Channel, 12-Roller
$1,035.00USD / Each