Ismatec® Pump Tubing, 2-Stop Color-Coded Autoanalysis/Microbore, Platinum-Cured Silicone
Designed for optimal operation in your Ismatec pumps
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • No leachable additives or plasticizers
Appearance: Opaque
Temperature: -70 to 52°C
Life at 100 rpm: 1000 hrs
The overall length of platinum-cured silicone 95602-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
$112.00USD / Pkg of 6
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm)
Availability Pricing
COMPARE
Orange/white 0.64
$112.00USD / Pkg of 6
COMPARE
Black/black 0.76
$112.00USD / Pkg of 6
COMPARE
Orange/orange 0.89
$112.00USD / Pkg of 6
COMPARE
White/white 1.02
$112.00USD / Pkg of 6
COMPARE
Red/red 1.14
$112.00USD / Pkg of 6
COMPARE
Gray/gray 1.3
$112.00USD / Pkg of 6
COMPARE
Yellow/yellow 1.42
$112.00USD / Pkg of 6
COMPARE
Yellow/blue 1.52
$112.00USD / Pkg of 6
COMPARE
Blue/blue 1.65
$112.00USD / Pkg of 6
COMPARE
Green/green 1.85
$112.00USD / Pkg of 6
COMPARE
Purple/purple 2.06
$112.00USD / Pkg of 6
COMPARE
Purple/black 2.29
$112.00USD / Pkg of 6
COMPARE
Purple/white 2.79
$112.00USD / Pkg of 6
Showing 1- 13 of 13