Ismatec® Pump Tubing, 2-Stop Color-Coded Autoanalysis/Microbore, Viton®

Designed for optimal operation in your Ismatec pumps
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Acid and solvent transfer; high-temperature work
Characteristics: Opaque black
Resin certification: Tubing is manufactured according to GMP
Temperature range: -8 to 400°F (-13 to 204°C)
Sterilize: At 480°F (249°C) for 16 hours in a circulating hot air oven

The overall length of Viton® 97628-series stop tubing is 12" (304 mm). The legacy Ismatec® stop tubing was 180 mm long.

Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
$136.00 - $140.00USD / Pkg of 12

15 variations of this product are available.

See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm)
Availability Pricing
COMPARE
Orange/yellow 0.51
$136.00USD / Pkg of 12
COMPARE
Orange/white 0.64
$140.00USD / Pkg of 12
COMPARE
Black/Black 0.76
$140.00USD / Pkg of 12
COMPARE
Orange/orange 0.89
$140.00USD / Pkg of 12
COMPARE
White/white 1.02
$140.00USD / Pkg of 12
COMPARE
Red/red 1.14
$140.00USD / Pkg of 12
COMPARE
Grey/grey 1.3
$136.00USD / Pkg of 12
COMPARE
Yellow/yellow 1.42
$140.00USD / Pkg of 12
COMPARE
Yellow/blue 1.52
$140.00USD / Pkg of 12
COMPARE
Blue/blue 1.65
$140.00USD / Pkg of 12
COMPARE
Green/green 1.85
$140.00USD / Pkg of 12
COMPARE
Purple/purple 2.06
$140.00USD / Pkg of 12
COMPARE
Purple/black 2.29
$140.00USD / Pkg of 12
COMPARE
Purple/orange 2.54
$140.00USD / Pkg of 12
COMPARE
Purple/white 2.79
$140.00USD / Pkg of 12
Showing 1- 15 of 15

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat