Ismatec® Pump Tubing, 2-Stop Microbore, Platinum-Cured Silicone

Excellent biocompatibility that meets multiple classifications
  • Genuine Ismatec® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Compatible with Masterflex® L/S® Multichannel pump head, small cartridges.

Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications

Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent

For custom tubing lengths and configurations, please contact our Product Applications Specialist.
$112.00 - $113.00USD / Pkg of 6

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing formulation Tubing ID (mm)
Availability Pricing
COMPARE
Orange/Orange Platinum-Cured Silicone 0.89
$112.00USD / Pkg of 6
COMPARE
Purple/Purple Platinum-Cured Silicone 2.06
$113.00USD / Pkg of 6
COMPARE
Purple/White Platinum-Cured Silicone 2.79
$113.00USD / Pkg of 6
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat