Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® S3™ E-LFL
Designed for optimal operation in your Ismatec pumps
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Longest tubing life of all Tygon tubing formulations
  • Broad chemical compatibility
  • Clear for easy flow monitoring
To complete your Ismatec® pumping system, order two-stop color-coded and extension tubing. Tubing comes in various formulations for almost any pumping application. The stops on the two-stop tubing are color-coded according to tubing ID. Use this tubing with any of the Ismatec® standard cartridge pumps and pump heads. Cartridges withstand pressures up to 45 psi (2.9 bar).
The overall length of E-LFL 96449-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
$117.00 - $131.00USD / Pkg of 12
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm) Description
Availability Pricing
COMPARE
Orange/red 0.19 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 0.19 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Orange/blue 0.25 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 0.25 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Orange/green 0.38 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 0.38 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Orange/yellow 0.51 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 0.51 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Orange/white 0.64 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 0.64 mm ID; 12/PK
$118.00USD / Pkg of 12
COMPARE
Black/black 0.76 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 0.76 mm ID; 12/PK
$117.00USD / Pkg of 12
COMPARE
Orange/orange 0.89 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 0.89 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
White/white 1.02 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 1.02 mm ID; 12/PK
$117.00USD / Pkg of 12
COMPARE
Red/red 1.14 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 1.14 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Red/Gray 1.22 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 1.22 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Yellow/Yellow 1.37 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 1.37 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Yellow/yellow 1.42 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 1.42 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Yellow/blue 1.52 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 1.52 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Blue/Blue 1.6 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 1.60 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Green/green 1.85 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 1.85 mm ID; 12/PK
$118.00USD / Pkg of 12
COMPARE
Purple/purple 2.06 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 2.06 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Purple/Black 2.2 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 2.20 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Purple/Orange 2.62 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 2.62 mm ID; 12/PK
$131.00USD / Pkg of 12
COMPARE
Purple/white 2.79 Pump Tubing, 2-Stop, Tygon® S3™ E-LFL, 2.79 mm ID; 12/PK
$131.00USD / Pkg of 12
Showing 1- 19 of 19