Ismatec® Pump Tubing, 3-Stop Color-Coded Autoanalysis/Microbore, Tygon® LMT-55
Designed for optimal operation in your Ismatec pumps
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
The overall length of LMT-55 97619-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
$94.00USD / Pkg of 12
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm)
Availability Pricing
COMPARE
Orange/black 0.13
$94.00USD / Pkg of 12
COMPARE
Orange/red 0.19
$94.00USD / Pkg of 12
COMPARE
Orange/blue 0.25
$94.00USD / Pkg of 12
COMPARE
Orange/green 0.38
$94.00USD / Pkg of 12
COMPARE
Green/yellow 0.44
$94.00USD / Pkg of 12
COMPARE
White/yellow 0.57
$94.00USD / Pkg of 12
COMPARE
Orange/white 0.64
$94.00USD / Pkg of 12
COMPARE
Black/Black 0.76
$94.00USD / Pkg of 12
COMPARE
Orange/orange 0.89
$94.00USD / Pkg of 12
COMPARE
White/black 0.95
$94.00USD / Pkg of 12
COMPARE
White/red 1.09
$94.00USD / Pkg of 12
COMPARE
Grey/grey 1.3
$94.00USD / Pkg of 12
COMPARE
Yellow/yellow 1.42
$94.00USD / Pkg of 12
COMPARE
Blue/green 1.75
$94.00USD / Pkg of 12
COMPARE
Green/green 1.85
$94.00USD / Pkg of 12
COMPARE
Purple/purple 2.06
$94.00USD / Pkg of 12
COMPARE
Purple/black 2.29
$94.00USD / Pkg of 12
COMPARE
Purple/orange 2.54
$94.00USD / Pkg of 12
COMPARE
Purple/white 2.79
$94.00USD / Pkg of 12
COMPARE
Black/white 3.17
$94.00USD / Pkg of 12
Showing 1 - 20 of 26