Ismatec® Pump Tubing, 3-Stop Color-Coded Autoanalysis/Microbore, Tygon® S3™ E-Food

Designed for optimal operation in your Ismatec pumps
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Specially designed for use with food applications
  • Extremely smooth bore
Appearance Clear Temperature:-66 to 135°C Life at 100 rpm:1000 hours Advantages:Ultra-smooth inner bore, excellent biocompatibility for sensitive applications, non-cytotoxic, non-pyrogenic, and non-hemolytic, low protein binding. Heat-sealable, bondable, sterile-weldable, and custom moldable for assemblies.
Application: Pharmaceutical, biotech, and general-purpose lab applications. Sterile filling and processing, cell harvest and media process systems, filtration, fermentation, cell and tissue culture transport
$71.50USD / Pkg of 12

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm)
Availability Pricing
COMPARE
Orange/blue 0.25
$71.50USD / Pkg of 12
COMPARE
Orange/yellow 0.51
$71.50USD / Pkg of 12
COMPARE
Orange/orange 0.89
$71.50USD / Pkg of 12
COMPARE
Red/red 1.14
$71.50USD / Pkg of 12
COMPARE
Yellow/yellow 1.42
$71.50USD / Pkg of 12
COMPARE
Purple/purple 2.06
$71.50USD / Pkg of 12
COMPARE
Purple/white 2.79
$71.50USD / Pkg of 12
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat