Ismatec® Pump Tubing, 3-Stop Microbore, Versilon™ 2001
Designed for optimal operation in your Ismatec pumps
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attached tubing stops give you two sections—when one section of tubing fatigues, a fresh section is ready to use. The stops on the three-stop tubing are color-coded according to tubing ID. Use three-stop tubing with any of the Ismatec® minicartridge pumps and pump heads. Minicartridges withstand pressures up to 22 psi.
The Tygon® Chemical 2001 tubing is clear in appearance is temperature resistant from -108°F to 135°F, and features excellent chemical compatibility to weak and medium acids and bases, salts, and alcohol.
The overall length of Tygon Chemical® 96446-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
$106.00USD / Pkg
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm) Description
Availability Pricing
COMPARE
Orange/green 0.38 Pump Tubing, 3-Stop, Tygon® Chemical 2001, 0.38 mm ID; 6/PK
$106.00USD / Pkg of 6
COMPARE
Orange/white 0.64 Pump Tubing, 3-Stop, Tygon® Chemical 2001, 0.64 mm ID; 6/PK
$106.00USD / Pkg
COMPARE
White/white 1.02 Pump Tubing, 3-Stop, Tygon® Chemical 2001, 1.02 mm ID; 6/PK
$106.00USD / Pkg
COMPARE
Yellow/blue 1.52 Pump Tubing, 3-Stop, Tygon® Chemical 2001, 1.52 mm ID; 6/PK
$106.00USD / Pkg
COMPARE
Purple/purple 2.06 Pump Tubing, 3-Stop, Tygon® Chemical 2001, 2.06 mm ID; 6/PK
$106.00USD / Pkg
COMPARE
Purple/white 2.79 Pump Tubing, 3-Stop, Tygon® Chemical 2001, 2.79 mm ID; 6/PK
$106.00USD / Pkg