Ismatec® Pump Tubing, Platinum-Cured Silicone
Replace worn tubing before it ruptures!
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Slightly greater clarity than peroxide-cured tubing
  • Smooth surfaces; lower protein binding levels
  • Fewer potential leachables
This translucent tubing is ideal for pharmaceutical, biotech, peristaltic pumping, and food applications. Platinum-cured silicone tubing has excellent heat resistance, low compression set, is odorless, and nontoxic for optimal performance.
$61.00 - $80.50USD / Pkg of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (mm) Tubing OD (mm) Wall Thickness (mm)
Availability Pricing
COMPARE
3.2 6.4 1.6
$61.00USD / Pkg of 1
COMPARE
6.4 9.6 1.6
$80.50USD / Pkg of 1