Ismatec® Microbore Pump Tubing, Puri-Flex®
Long life tubing for pharma and bioprocess applications
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: General-purpose lab use requiring high performance and long life
Characteristics: Exceptional tensile and tear strength, good biocompatibility, low gas permeability, smooth surface, translucent
Certifications: FDA-compliant (21 CFR 177.2600), meets USP Class VI requirements, manufactured according to GMP
Temperature range: –58 to 275°F (–50 to 135°C)
Sterilize: Autoclave, ethylene oxide, or gamma irradiation
$67.50 - $69.00USD / Pkg of 1
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
C/L Masterflex tubing size
Availability Pricing
COMPARE
10
$69.00USD / Pkg of 1
COMPARE
12
$69.00USD / Pkg of 1
COMPARE
14
$69.00USD / Pkg of 1
COMPARE
16
$69.00USD / Pkg of 1
COMPARE
18
$69.00USD / Pkg of 1
COMPARE
22
$67.50USD / Pkg of 1
COMPARE
24
$69.00USD / Pkg of 1
COMPARE
26
$69.00USD / Pkg of 1
COMPARE
28
$69.00USD / Pkg of 1
COMPARE
30
$67.50USD / Pkg of 1
COMPARE
32
$69.00USD / Pkg of 1
COMPARE
34
$69.00USD / Pkg of 1
COMPARE
36
$69.00USD / Pkg of 1
COMPARE
38
$67.50USD / Pkg of 1
COMPARE
40
$69.00USD / Pkg of 1
COMPARE
42
$69.00USD / Pkg of 1
COMPARE
44
$67.50USD / Pkg of 1
COMPARE
46
$67.50USD / Pkg of 1
COMPARE
48
$69.00USD / Pkg of 1
Showing 1- 19 of 19