Ismatec® Pump Tubing, Solvent/Hydrocarbon
Specially formulated to transport hydrocarbons, petroleum products and distillates
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Resistant to ozone and UV light
Resists to cracking, swelling, hardening
Ideal for fuels, distillates, lubricants, and glycol-based coolants
$54.50 - $56.50USD / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
C/L Masterflex tubing size Description
Availability Pricing
COMPARE
22 Extension Tubing, Solvent/Hydrocarbon 0.64 mm ID, 50 ft/pk
$54.50USD / Pkg of 1
COMPARE
24 Extension Tubing, Solvent/Hydrocarbon, 0.76 mm ID, 50 ft/pk
$54.50USD / Pkg of 1
COMPARE
28 Extension Tubing, Solvent/Hydrocarbon, 1.02 mm ID, 50 ft/pk
$56.50USD / Pkg of 1
COMPARE
38 Extension Tubing, Solvent/Hydrocarbon, 1.65 mm ID, 50 ft/pk
$56.50USD / Pkg of 1
COMPARE
44 Extension Tubing, Solvent/Hydrocarbon, 2.29 mm ID, 50 ft/pk
$54.50USD / Pkg of 1
COMPARE
46 Extension Tubing, Solvent/Hydrocarbon, 2.54 mm ID, 50 ft/pk
$54.50USD / Pkg of 1