Ismatec® Pump Tubing, Tygon® E-LFL
Longest life of all Tygon® formulations
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: General laboratory use
Characteristics: Long life, won’t age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic, transparent
$162.00USD / Pkg of 1
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
C/L Masterflex tubing size
Availability Pricing
COMPARE
10
$162.00USD / Pkg of 1
COMPARE
12
$162.00USD / Pkg of 1
COMPARE
14
$162.00USD / Pkg of 1
COMPARE
18
$162.00USD / Pkg of 1
COMPARE
22
$162.00USD / Pkg of 1
COMPARE
24
$162.00USD / Pkg of 1
COMPARE
26
$162.00USD / Pkg of 1
COMPARE
28
$162.00USD / Pkg of 1
COMPARE
30
$162.00USD / Pkg of 1
COMPARE
31
$162.00USD / Pkg of 1
COMPARE
33
$162.00USD / Pkg of 1
COMPARE
34
$162.00USD / Pkg of 1
COMPARE
36
$162.00USD / Pkg of 1
COMPARE
37
$162.00USD / Pkg of 1
COMPARE
40
$162.00USD / Pkg of 1
COMPARE
42
$162.00USD / Pkg of 1
COMPARE
43
$162.00USD / Pkg of 1
COMPARE
47
$162.00USD / Pkg of 1
COMPARE
48
$162.00USD / Pkg of 1
Showing 1- 19 of 19