Ismatec® Reglo Independent Channel Control (ICC) Panel-Mount Peristaltic Pumps
Independently control flow and direction of each channel via keypad or your PC
  • Flow rate 0.0002 to 35 mL/min per channel - flow rate depends on drive rpm, rollers, and tubing size
  • USB interface for quick connection to a PC
  • Flexibility of bidirectional flow in each channel
These multi-channel peristaltic pumps let you individually control the flow and direction of each channel via your computer—expanding your pumping capabilities with a single compact unit! Pump delivers continuous pumping or precision dispensing with the capability of bidirectional flow in each channel. Plus, independent channel calibration minimizes tube-to-tube differences, resulting in high calibration accuracy.
The panel-mount Reglo ICC pump offers many of the same features as on the standard ICC console pump. The only difference is the touchpad and LCD display is not included. Instead, easily integrate these into your system using the provided connections.
Order the three-stop tubing size and formulation to suit your application needs (sold separately). More than 100 options exist — the most popular sizes and formulations can be found under Accessories below
Ismatec software downloads can be found here.
$2,208.00 - $2,873.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate Per Channel (mL/min) Number of Channels Number of Rollers
Availability Pricing
COMPARE
35 2 8
$2,208.00USD / Each
COMPARE
35 3 8
$2,493.00USD / Each
COMPARE
35 4 8
$2,778.00USD / Each
COMPARE
24 4 12
$2,873.00USD / Each