This product is now discontinued, View Related Products
Isocratic Dispensing Piston Pumps
Deliver precise volumes at pressures ut to 6000 psi
  • Ideal for use in HPLC applications
  • Pulse dampener significantly reduces pulsation with low dead volume
Use this single-head pump for isocratic, metering, research and development and quality assurance applications. The low dead volume pulse damper and rapid refill in the pump make the flow virtually pulse-free. Easy solvent changes are accomplished via the one-button purging/priming feature.
Linear cam and pressure compensation provide accurate gradients in highor low-pressure modes. Chemical-resistant keypad and digital LED display allow flow adjustments in 0.01-mL increments. Other features include RS-232 serial communication port, run/stop inputs, remote frequency and voltage controls, dual-voltage capabilities, and real-time pressure monitoring.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Max Pressure (PSI) Wetted Materials
Availability Pricing
COMPARE
10 6000 316 SS, fluorocarbon, ruby, sapphire