This product is now discontinued, View Related Products
Isolation Barrier Gowns
Disposable, low-cost gowns provide full frontal protection from particulates or light splashes
Lightweight breathable gowns feature ties in back of neck and at waist for a secure, comfortable fit. Elastic cuffs at wrists keep the sleeves firmly in place and gather softly under gloves. Select 100% spunbound polypropylene blue gowns for durable, particulate protection. Select polyethylene-coated polypropylene yellow gowns for added protection from water based liquids and certain chemicals. Sold in case of 50.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Length (in) Size
Availability Pricing
Total Source APP0270-BLUE Isolation barrier gowns, polypropylene, case of 50 EW-86566-00 Mfr # APP0270-BLUE
COMPARE
Blue 45.5 Universal
Call For Price
Total Source APP0278-YELLOW Isolation barrier gowns, polyethylene-coated polypropylene, case of 50 EW-86566-10 Mfr # APP0278-YELLOW
COMPARE
Yellow 45.5 Universal