John Guest acetal push-to-connect end stops
  • Meet NSF and FDA compliance for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
$19.70 - $26.25USD / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Push-to-Connect Tube OD (port 1) (in)
Availability Pricing
John Guest Push-to-connect end stops, 1/4", 10/pack
Buy More Save More
EW-34007-18 Mfr # PI4608S
COMPARE
Acetal 1/4
$19.70USD / Pkg of 10
John Guest Push-to-connect end stops, 3/8", 10/pack
Buy More Save More
EW-34007-19 Mfr # PI4612S
COMPARE
Acetal 3/8
$26.25USD / Pkg of 10