John Guest Acetal Push-To-Connect Reducing Tee Adapter Fittings
No need to buy two fitting halves
  • Meets NSF and FDA compliance for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
$70.50 - $74.00USD / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Push-to-Connect Tube OD (port 1) (in) Push-to-Connect Tube OD (port 2) (in) Push-to-Connect Tube OD (port 3) (in)
Availability Pricing
John Guest Push-To-Connect Fitting, Reducing Tee Adapter, Acetal, 1/2" OD x 3/8" OD; 10/Pk
Buy More Save More
EW-34006-01 Mfr # PI301612S
COMPARE
1/2 1/2 3/8
$70.50USD / Pkg of 10
John Guest Push-To-Connect Fitting, Reducing Tee Adapter, Acetal, 3/8" OD x 1/4" OD; 10/Pk
Buy More Save More
EW-34006-47 Mfr # PI301208S
COMPARE
3/8 3/8 1/4
$74.00USD / Pkg of 10