John Guest acetal push-to-connect to hose barb elbow swivel adapters
  • Meet NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
$26.25 - $39.25USD / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
John Guest Push-to-connect stem-to-hose elbow adapters, 1/4" x 1/4", 10/pack
Buy More Save More
EW-34006-84 Mfr # PI290808S
COMPARE
1/4 Acetal
$26.25USD / Pkg of 10
John Guest Push-to-connect stem-to-hose elbow adapters, 3/8" x 1/4", 10/pack
Buy More Save More
EW-34006-85 Mfr # PI291208S
COMPARE
1/4 Acetal
$32.75USD / Pkg of 10
John Guest Push-to-connect stem-to-hose elbow adapters, 5/16" x 5/16", 10/pack
Buy More Save More
EW-34007-12 Mfr # PM290808S
COMPARE
5/16 Acetal
$30.75USD / Pkg of 10
John Guest Push-to-connect stem-to-hose elbow adapters, 1/4" x 5/16", 10/pack
Buy More Save More
EW-34007-13 Mfr # PI290810S
COMPARE
5/16 Acetal
$31.75USD / Pkg of 10
John Guest Push-to-connect stem-to-hose elbow adapters, 3/8" x 5/16", 10/pack
Buy More Save More
EW-34007-15 Mfr # PI291210S
COMPARE
5/16 Acetal
$39.25USD / Pkg of 10