John Guest acetal push-to-connect to threaded elbow adapters
  • Meet NSF and FDA compliance for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
$31.75 - $46.25USD / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Push-to-Connect Tube OD (port 1) (in) Thread size NPT(M) (port 2) (in)
Availability Pricing
John Guest Push-to-connect fixed threaded male elbow adapters, 1/4" x 1/8" NPTM, 10/pack
Buy More Save More
EW-34007-21 Mfr # PI480821S
COMPARE
Acetal 1/4 1/8
$31.75USD / Pkg of 10
John Guest Push-to-connect fixed threaded male elbow adapters, 1/4" x 1/4" NPTM, 10/pack
Buy More Save More
EW-34007-22 Mfr # PI480822S
COMPARE
Acetal 1/4 1/4
$35.75USD / Pkg of 10
John Guest Push-to-connect fixed threaded male elbow adapters, 1/4" x 3/8" NPTM, 10/pack
Buy More Save More
EW-34007-23 Mfr # PI480823S
COMPARE
Acetal 1/4 3/8
$39.25USD / Pkg of 10
John Guest Push-to-connect fixed threaded male elbow adapters, 3/8" x 3/8" NPTM, 10/pack
Buy More Save More
EW-34007-25 Mfr # PI481223S
COMPARE
Acetal 3/8 3/8
$46.25USD / Pkg of 10