John Guest Acetal Push-To-Connect Male Threaded Adapter Fittings
No need to buy two fitting halves
  • Meets NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
  • A single fitting offers secure connections
  • and quick disconnection
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
$11.00 - $29.25USD / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Push-to-Connect Tube OD (port 1) (in) Thread size NPT(M) (port 2) (in)
Availability Pricing
John Guest Push-To-Connect Fitting, Male Threaded Adapter, Acetal, 1/4" OD x 1/8" NPT(M); 10/Pk
Buy More Save More
EW-34006-66 Mfr # PI050821S
COMPARE
Acetal 1/4 1/8
$11.10USD / Pkg of 10
John Guest Push-To-Connect Fitting, Male Threaded Adapter, Acetal, 1/4" OD x 1/4" NPT(M); 10/Pk
Buy More Save More
EW-34006-67 Mfr # PI050822S
COMPARE
Acetal 1/4 1/4
$11.40USD / Pkg of 10
John Guest Push-To-Connect Fitting, Male Threaded Adapter, Acetal, 3/8" OD x 1/4" NPT(M); 10/Pk
Buy More Save More
EW-34006-68 Mfr # PI051222S
COMPARE
Acetal 3/8 1/4
$16.60USD / Pkg of 10
John Guest Push-To-Connect Fitting, Male Threaded Adapter, Acetal, 3/8" OD x 3/8" NPT(M); 10/Pk
Buy More Save More
EW-34006-69 Mfr # PI051223S
COMPARE
Acetal 3/8 3/8
$16.10USD / Pkg of 10
John Guest Push-To-Connect Fitting, Male Threaded Adapter, Acetal, 1/2" OD x 3/8" NPT(M); 10/Pk
Buy More Save More
EW-34006-70 Mfr # PI051623S
COMPARE
Acetal 1/2 3/8
$28.25USD / Pkg of 10
John Guest Push-To-Connect Fitting, Male Threaded Adapter, Acetal, 1/2" OD x 1/2" NPT(M); 10/Pk
Buy More Save More
EW-34006-71 Mfr # PI051624S
COMPARE
Acetal 1/2 1/2
$29.25USD / Pkg of 10
John Guest Push-To-Connect Fitting, Male Threaded Adapter, Acetal, 3/16" OD x 1/8" NPT(M); 10/Pk
Buy More Save More
EW-34006-86 Mfr # PI050621S
COMPARE
Acetal 3/16 3/8
$22.50USD / Pkg of 10
John Guest Push-To-Connect Fitting, Male Threaded Adapter, Acetal, 5/32" OD x 1/8" NPT(M); 10/Pk
Buy More Save More
EW-34007-27 Mfr # PM050421S
COMPARE
Acetal 5/32 1/8
$11.40USD / Pkg of 10
John Guest Push-To-Connect Fitting, Male Threaded Adapter, Acetal, 5/32" OD x 1/4" NPT(M); 10/Pk
Buy More Save More
EW-34007-28 Mfr # PM050422S
COMPARE
Acetal 5/32 1/4
$11.00USD / Pkg of 10
John Guest Push-To-Connect Fitting, Male Threaded Adapter, Acetal, 5/16" OD x 1/8" NPT(M); 10/Pk
Buy More Save More
EW-34007-29 Mfr # PM050821S
COMPARE
Acetal 5/16 1/8
$11.00USD / Pkg of 10
John Guest Push-To-Connect Fitting, Male Threaded Adapter, Acetal, 5/16" OD x 1/4" NPT(M); 10/Pk
Buy More Save More
EW-34007-30 Mfr # PM050822S
COMPARE
Acetal 5/16 1/4
$11.00USD / Pkg of 10
John Guest Push-To-Connect Fitting, Male Threaded Adapter, Acetal, 5/16" OD x 3/8" NPT(M); 10/Pk
Buy More Save More
EW-34007-31 Mfr # PM050823S
COMPARE
Acetal 5/16 3/8
$16.60USD / Pkg of 10
Showing 1- 12 of 12