John Guest Acetal push-to-connect tube-to-hose barb adapters
No need to buy two fitting halves
  • Meet NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
$17.90 - $44.75USD / Pkg of 10
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material Push-to-Connect Tube OD (port 1) (in)
Availability Pricing
John Guest Push-to-connect stem-to-hose adapters, 1/4" x 1/4", 10/pack
Buy More Save More
EW-34006-79 Mfr # PI250808S
COMPARE
1/4 Acetal 1/4
$18.40USD / Pkg of 10
John Guest Push-to-connect stem-to-hose adapters, 3/8" x 1/4", 10/pack
Buy More Save More
EW-34006-80 Mfr # PI251208S
COMPARE
1/4 Acetal 3/8
$28.75USD / Pkg of 10
John Guest Push-to-connect stem-to-hose adapters, 3/8" x 3/8", 10/pack
Buy More Save More
EW-34006-81 Mfr # PI251212S
COMPARE
3/8 Acetal 3/8
$32.25USD / Pkg of 10
John Guest Push-to-connect stem-to-hose adapters, 1/2" x 3/8", 10/pack
Buy More Save More
EW-34006-82 Mfr # PI251612S
COMPARE
3/8 Acetal 1/2
$33.75USD / Pkg of 10
John Guest Push-to-connect stem-to-hose adapters, 1/2" x 1/2", 10/pack
Buy More Save More
EW-34006-83 Mfr # PI251616S
COMPARE
1/2 Acetal 1/2
$44.75USD / Pkg of 10
John Guest Push-to-connect stem-to-hose adapters, 1/4" x 3/16", 10/pack
Buy More Save More
EW-34007-04 Mfr # PI250806S
COMPARE
3/16 Acetal 1/4
$18.40USD / Pkg of 10
John Guest Push-to-connect stem-to-hose adapters, 1/4" x 5/16", 10/pack
Buy More Save More
EW-34007-05 Mfr # PI250810S
COMPARE
5/16 Acetal 1/4
$17.90USD / Pkg of 10
John Guest Push-to-connect stem-to-hose adapters, 5/16" x 3/16", 10/pack
Buy More Save More
EW-34007-06 Mfr # PI251006S
COMPARE
3/16 Acetal 5/16
$18.40USD / Pkg of 10
John Guest Push-to-connect stem-to-hose adapters, 5/16" x 1/4", 10/pack
Buy More Save More
EW-34007-07 Mfr # PI251008S
COMPARE
1/4 Acetal 5/16
$20.25USD / Pkg of 10
John Guest Push-to-connect stem-to-hose adapters, 5/16" x 3/8", 10/pack
Buy More Save More
EW-34007-08 Mfr # PI251012S
COMPARE
3/8 Acetal 5/16
$27.75USD / Pkg of 10
John Guest Push-to-connect stem-to-hose adapters, 3/8" x 5/16", 10/pack
Buy More Save More
EW-34007-09 Mfr # PI251210S
COMPARE
5/16 Acetal 3/8
$29.25USD / Pkg of 10
John Guest Push-to-connect stem-to-hose adapters, 3/8" x 1/2", 10/pack
Buy More Save More
EW-34007-11 Mfr # PI251216S
COMPARE
1/2 Acetal 3/8
$39.25USD / Pkg of 10
John Guest Push-to-connect stem-to-hose adapters, 5/16" x 5/16", 10/pack
Buy More Save More
EW-34007-37 Mfr # PM250808S
COMPARE
5/16 Acetal 5/16
$20.25USD / Pkg of 10
Showing 1- 13 of 13