John Guest acetal push-to-connect union T-connectors
  • Meet NSF and FDA compliance for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth— tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
$36.75 - $61.00USD / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Push-to-Connect Tube OD (port 1) (in) Push-to-Connect Tube OD (port 2) (in)
Availability Pricing
John Guest Tee Fittings, for 1/4" OD Tubing, Acetal, 10/Pk
Buy More Save More
EW-34006-11 Mfr # PI0208S
COMPARE
Acetal 1/4 1/4
$41.75USD / Pkg of 10
John Guest Tee Fittings, for 3/8" OD Tubing, Acetal, 10/Pk
Buy More Save More
EW-34006-12 Mfr # PI0212S
COMPARE
Acetal 3/8 3/8
$51.50USD / Pkg of 10
John Guest Tee Fittings, for 1/2" OD Tubing, Acetal, 10/Pk
Buy More Save More
EW-34006-13 Mfr # PI0216S
COMPARE
Acetal 1/2 1/2
$61.00USD / Pkg of 10
John Guest Tee Fittings, for 3/16" OD Tubing, Acetal, 10/Pk
Buy More Save More
EW-34006-30 Mfr # PI0206S
COMPARE
Acetal 3/16 3/16
$37.00USD / Pkg of 10
John Guest Tee Fittings, for 5/32" OD Tubing, Acetal, 10/Pk
Buy More Save More
EW-34006-53 Mfr # PM0204S
COMPARE
Acetal 5/32 5/32
$39.25USD / Pkg of 10
John Guest Tee Fittings, for 5/16" OD Tubing, Acetal, 10/Pk
Buy More Save More
EW-34006-54 Mfr # PM0208S
COMPARE
Acetal 5/16 5/16
$41.50USD / Pkg of 10
John Guest Push-to-connect union tees, polypropylene, 1/4", 10/pack
Always in Stock
EW-34007-46 Mfr # PP0208W
COMPARE
Polypropylene 1/4 1/4
$36.75USD / Pkg of 10
John Guest Push-to-connect union tees, polypropylene, 3/8", 10/pack EW-34007-47 Mfr # PP0212W
COMPARE
Polypropylene 3/8 3/8
$46.75USD / Pkg of 10
Showing 1- 8 of 8