John Guest Polypropylene Push-To-Connect Elbow Adapter Fittings
Quick, leakproof connection
  • Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications, and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
Adapters let you permanently modify basic fittings for customized installations.
$29.75 - $45.25USD / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 2) (in) Fitting Material Push-to-Connect Tube OD (port 1) (in)
Availability Pricing
John Guest Push-To-Connect Fitting, Elbow Adapter, Polypropylene, 1/4" OD x 1/4" OD; 10/Pk EW-34007-58 Mfr # PP220808W-US
COMPARE
1/4 Polypropylene 1/4
$29.75USD / Pkg of 10
John Guest Push-To-Connect Fitting, Elbow Adapter, Polypropylene, 3/8" OD x 1/4" OD; 10/Pk EW-34007-59 Mfr # PP221208W
COMPARE
1/4 Polypropylene 3/8
$40.75USD / Pkg of 10
John Guest Push-To-Connect Fitting, Elbow Adapter, Polypropylene, 3/8" OD x 3/8" OD; 10/Pk EW-34007-60 Mfr # PP221212W-US
COMPARE
3/8 Polypropylene 3/8
$45.25USD / Pkg of 10