John Guest Push-to-Connect Plug Inserts, Acetal
No need to buy two fitting halves
  • Meet NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
$7.30 - $19.90USD / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Push-to-Connect Tube OD (port 1) (in)
Availability Pricing
John Guest  Push-to-Connect Plug Insert, Acetal, 1/4" OD; 10/Pk
Buy More Save More
EW-34006-73 Mfr # PI0808S
COMPARE
Acetal 1/4
$9.50USD / Pkg of 10
John Guest  Push-to-Connect Plug Insert, Acetal, 3/8" OD; 10/Pk
Buy More Save More
EW-34006-74 Mfr # PI0812S
COMPARE
Acetal 3/8
$17.10USD / Pkg of 10
John Guest  Push-to-Connect Plug Insert, Acetal, 1/2" OD; 10/Pk
Buy More Save More
EW-34006-75 Mfr # PI0816S
COMPARE
Acetal 1/2
$19.90USD / Pkg of 10
John Guest  Push-to-Connect Plug Insert, Acetal, 3/16" OD; 10/Pk
Buy More Save More
EW-34006-93 Mfr # PI0806S
COMPARE
Acetal 3/16
$7.30USD / Pkg of 10
John Guest  Push-to-Connect Plug Insert, Acetal, 5/32" OD; 10/Pk
Buy More Save More
EW-34007-32 Mfr # PM0804S
COMPARE
Acetal 5/32
$7.60USD / Pkg of 10
John Guest  Push-to-Connect Plug Insert, Acetal, 5/16" OD; 10/Pk
Buy More Save More
EW-34007-33 Mfr # PM0808S
COMPARE
Acetal 5/16
$11.00USD / Pkg of 10