John Guest push-to-connect to male NPT adapters
No need to buy two fitting halves
  • Meet NSF and FDA compliance for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth— tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
$17.10 - $58.00USD / Pkg of 10
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Push-to-Connect Tube OD (port 1) (in) Thread size NPT(M) (port 2) (in)
Availability Pricing
John Guest Male Pipe Adapters, 1/4 X 1/8, 10 Per Pack
Buy More Save More
EW-34006-02 Mfr # PI010821S
COMPARE
Acetal 1/4 1/8
$19.90USD / Pkg of 10
John Guest Male Pipe Adapters, 1/4 X 1/4, 10 Per Pack
Buy More Save More
EW-34006-03 Mfr # PI010822S
COMPARE
Acetal 1/4 1/4
$23.50USD / Pkg of 10
John Guest Male Pipe Adapters, 3/8 X 1/8, 10 Per Pack
Buy More Save More
EW-34006-04 Mfr # PI011221S
COMPARE
Acetal 3/8 1/8
$42.75USD / Pkg of 10
John Guest Male Pipe Adapters, 3/8 X 1/4, 10 Per Pack
Buy More Save More
EW-34006-05 Mfr # PI011222S
COMPARE
Acetal 3/8 1/4
$33.75USD / Pkg of 10
John Guest Threaded Adapters, 3/8" OD to 3/8" NPT(M), Acetal, 10/Pk
Buy More Save More
EW-34006-06 Mfr # PI011223S
COMPARE
Acetal 3/8 3/8
$39.25USD / Pkg of 10
John Guest Male Pipe Adapters, 3/8 X 1/2, 10 Per Pack
Buy More Save More
EW-34006-07 Mfr # PI011224S
COMPARE
Acetal 3/8 1/2
$57.00USD / Pkg of 10
John Guest Male Pipe Adapters, 1/2 X 3/8, 10 Per Pack
Buy More Save More
EW-34006-08 Mfr # PI011623S
COMPARE
Acetal 1/2 3/8
$49.00USD / Pkg of 10
John Guest Male Pipe Adapters, 1/2 X 1/2, 10 Per Pack
Buy More Save More
EW-34006-09 Mfr # PI011624S
COMPARE
Acetal 1/2 1/2
$58.00USD / Pkg of 10
John Guest Threaded Adapters, 3/16" OD to 1/8" NPT(M), Acetal, 10/Pk
Buy More Save More
EW-34006-28 Mfr # PI010621S
COMPARE
Acetal 3/16 1/8
$18.40USD / Pkg of 10
John Guest Threaded Adapters, 1/4" OD to 3/8" NPT(M), Acetal, 10/Pk
Buy More Save More
EW-34006-29 Mfr # PI010823S
COMPARE
Acetal 1/4 3/8
$31.75USD / Pkg of 10
John Guest Threaded Adapters, 5/32" OD to 1/8" NPT(M), Acetal, 10/Pk
Buy More Save More
EW-34006-48 Mfr # PM010421S
COMPARE
Acetal 5/32 1/8
$17.10USD / Pkg of 10
John Guest Threaded Adapters, 5/32" OD to 1/4" NPT(M), Acetal, 10/Pk
Buy More Save More
EW-34006-49 Mfr # PM010422S
COMPARE
Acetal 5/32 1/4
$20.50USD / Pkg of 10
John Guest Threaded Adapters, 5/16" OD to 1/8" NPT(M), Acetal, 10/Pk
Buy More Save More
EW-34006-50 Mfr # PM010821S
COMPARE
Acetal 5/16 1/8
$26.75USD / Pkg of 10
John Guest Threaded Adapters, 5/16" OD to 1/4" NPT(M), Acetal, 10/Pk
Buy More Save More
EW-34006-51 Mfr # PM010822S
COMPARE
Acetal 5/16 1/4
$24.00USD / Pkg of 10
John Guest Threaded Adapters, 5/16" OD to 3/8" NPT(M), Acetal, 10/Pk
Buy More Save More
EW-34006-52 Mfr # PM010823S
COMPARE
Acetal 5/16 3/8
$35.75USD / Pkg of 10
John Guest Push-to-connect male adapter, polypropylene, 1/4" x 1/8" NPTM, 10/pack EW-34007-42 Mfr # PP010821W
COMPARE
Polypropylene 1/4 1/8
$18.40USD / Pkg of 10
John Guest Push-to-connect male adapter, polypropylene, 1/4" x 1/4" NPTM, 10/pack EW-34007-43 Mfr # PP010822W
COMPARE
Polypropylene 1/4 1/4
$19.90USD / Pkg of 10
John Guest Push-to-connect male adapter, polypropylene, 3/8" x 1/4" NPTM, 10/pack EW-34007-44 Mfr # PP011222W
COMPARE
Polypropylene 3/8 1/4
$33.75USD / Pkg of 10
John Guest Push-to-connect male adapter, polypropylene, 3/8" x 3/8" NPTM, 10/pack EW-34007-45 Mfr # PP011223W
COMPARE
Polypropylene 3/8 3/8
$35.50USD / Pkg of 10
Showing 1- 19 of 19