Justrite Sure-Grip® EX Compact Flammable Cabinets
Flammable compact cabinets are ideal for benchtop use
  • Meets OSHA and NFPA standards
Use these 18-gauge steel constructed cabinets to safely store flammable or combustible liquids. Fire-resistant design includes double-walled steel with 1-1/2-in (38-mm) of insulating air space and 3-point stainless steel bullet latch for positive, failsafe door closure.
Cabinets come equipped with one adjustable SpillSlope® safety shelf that directs spills to leakproof 2” (51 mm) sump. Other features include dual vents with flame arresters, leveling feet, and grounding connector. The cabinets can easily fit under an existing countertop or use as standalone units.
$1,021.00 - $1,552.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Self-Closing
Availability Pricing
Justrite 890420 Sure-Grip® EX Countertop Flammable Cabinet, Self Close, 1 Door, 4 Gal EW-81790-20 Mfr # 890420
COMPARE
4 Yes
$1,333.00USD / Each
Justrite 890400 Sure-Grip® EX Countertop Flammable Cabinet, Manual Close, 1 Door,  4 Gal EW-81790-21 Mfr # 890400
COMPARE
4 No
$1,021.00USD / Each
Justrite 891220 Sure-Grip® EX Compact Flammable Cabinet, Self Close, 1 Door, 12 Gal EW-81790-22 Mfr # 891220
COMPARE
12 Yes
$1,552.00USD / Each
Justrite 891200 Sure-Grip® EX Compact Flammable Cabinet, Manual Close, 1 Door, 12 Gal EW-81790-23 Mfr # 891200
COMPARE
12 No
$1,239.00USD / Each