Justrite Quick-Disconnect HPLC Poly Manifolds
A cost-effective way to collect HPLC waste
These polypropylene manifolds accept up to six waste lines from HPLC processes to be fed into a single intake fitting. Manifolds have six 1/4-28 ports, one 3/8" NPT port (all come with caps for unused ports), and one quick-disconnect port in either 316 stainless steel or polypropylene.
$323.50 - $596.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Justrite 28177 HPLC Poly Manifold w/Stainless Steel Quick Disconnect for Safety Disposal Can EW-86394-27 Mfr # 28177
COMPARE
HPLC Poly Manifold w/stainless quick disconnect
$596.50USD / Each
Justrite 28178 HPLC Poly Manifold w/Polypropylene Quick Disconnect for Safety Disposal Can EW-86394-29 Mfr # 28178
COMPARE
HPLC Poly Manifold w/polypropylene quick disconnect
$350.50USD / Each
Justrite 28179 HPLC Poly Manifold w/Stainless Steel Stacker Connector for up to 12 Waste Lines EW-86394-31 Mfr # 28179
COMPARE
HPLC Poly Manifold w/stainless \"stacker\" connector
$323.50USD / Each