KIMAX® Class A Volumetric Flasks
Get accurate dilutions
  • Volumetric flask features single graduation ring blasted on the neck
  • Constructed of low extractable borosilicate glass
  • Autoclavable
These Class A volumetric flasks are calibrated "To Contain" and have marking areas flasks. The 1-mL and 2-mL flasks are of a test tube shape, all other sizes are conventional flasks. The 5-mL and larger flasks meet Class A ASTM Specification E288 unserialized specifications. The 1- and 2-mL sizes are test tube-shaped and are calibrated to E237 tolerances.
$496.00 - $1,117.00USD / Case of 12
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
DWK Life Sciences (Kimble) Class A Volumetric Glass Flask, Test Tube Shaped, 1 mL; 12/CS EW-34528-00 Mfr # 28014-1
COMPARE
1
$590.00USD / Case of 12
DWK Life Sciences (Kimble) Class A Volumetric Glass Flask, Test Tube Shaped, 2 mL; 12/CS EW-34528-02 Mfr # 28014-2
COMPARE
2
$631.00USD / Case of 12
DWK Life Sciences (Kimble) Class A Volumetric Glass Flask, 5 mL; 12/CS EW-34528-04 Mfr # 28014-5
COMPARE
5
$496.00USD / Case of 12
DWK Life Sciences (Kimble) Class A Volumetric Glass Flask, 10 mL; 12/CS EW-34528-06 Mfr # 28014-10
COMPARE
10
$590.00USD / Case of 12
DWK Life Sciences (Kimble) Class A Volumetric Glass Flask, 25 mL; 12/CS EW-34528-08 Mfr # 28014-25
COMPARE
25
$637.50USD / Case of 12
DWK Life Sciences (Kimble) Class A Volumetric Glass Flask, 50 mL; 12/CS EW-34528-10 Mfr # 28014-50
COMPARE
50
$677.50USD / Case of 12
DWK Life Sciences (Kimble) Class A Volumetric Glass Flask, 100 mL; 12/CS EW-34528-12 Mfr # 28014-100
COMPARE
100
$724.00USD / Case of 12
DWK Life Sciences (Kimble) Class A Volumetric Glass Flask, 200 mL; 12/CS EW-34528-14 Mfr # 28014-200
COMPARE
200
$828.50USD / Case of 12
DWK Life Sciences (Kimble) Class A Volumetric Glass Flask, 250 mL; 12/CS EW-34528-16 Mfr # 28014-250
COMPARE
250
$897.50USD / Case of 12
DWK Life Sciences (Kimble) Class A Volumetric Glass Flask, 500 mL; 12/CS EW-34528-18 Mfr # 28014-500
COMPARE
500
$1,117.00USD / Case of 12
DWK Life Sciences (Kimble) Class A Volumetric Glass Flask, 1000 mL; 6/CS EW-34528-20 Mfr # 28014-1000
COMPARE
1000
$677.50USD / Case of 6
DWK Life Sciences (Kimble) Class A Volumetric Glass Flask, 2000 mL; 4/CS EW-34528-22 Mfr # 28014-2000
COMPARE
2000
$572.50USD / Case of 4
Showing 1- 12 of 12