KIMAX Shellbach Burettes (Burets)
Significantly increases your visibility and reading accuracy
  • Maintain fluid integrity within burette (buret) with dust cap
  • Designed from ASTM Specification E287 requirements
Shellbach straight-bore burettes (burets) are designed with a blue line on a broader white band running vertically behind the scale to create an optical effect which makes the meniscus and line to appear to form a point. Read meniscus at the narrowest point of the blue line for best results. Scale is durable black ceramic enamel.
$201.00 - $204.50USD / Case of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Tolerance
Availability Pricing
DWK Life Sciences (Kimble) Kimax Shellback burettes with dust cap 25 ml EW-34542-50 Mfr # 17150F-25
COMPARE
25 0.06
$201.00USD / Case of 1
DWK Life Sciences (Kimble) Kimax Shellback burettes with dust cap 50 ml EW-34542-52 Mfr # 17150F-50
COMPARE
50 0.1
$204.50USD / Case of 1