This product is now discontinued, View Related Products
Kinesis KX Hydrophilic PTFE Syringe Filters with Prefilter
Hydrophilic PTFE membranes exhibit excellent chemical resistance and are high protein binding
  • Improve removal of larger and unwanted buildup with a two-stage filter
  • Increase flow rate with a less clogged and longer life membrane
  • Leak-free Luer-Lok® and Luer-slip connections
  • Color-coded filters for easy organization
Prepare reliable aqueous, organic, and biomolecular samples with membrane filtration and a prefilter (depth filter) inside a polypropylene (PP) housing. The prefilter adds a filtration stage to remove larger particles which create unwanted buildup. Useful with solutions with a high viscosity like oils or high particulate samples.
Each housing comes with a color-coded retainer ring seal to prevent leaks and sample loss, and the color helps to visually show you the membrane type inside the housing. Syringe filters have Luer-Lok and Luer-slip connections for easy, quick and tight connections to tubing, syringes, or needles. Filter packs are available in a resealable storage container to prevent contamination during multiple opening and closings.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Diameter (mm) Pore Size (µm) Description
Availability Pricing
COMPARE
0.22 Syringe Filter w/ Prefilter, Hydrophilic PTFE, 13 mm dia., 0.22 µm; 100/pk
Call For Price
COMPARE
0.45 Syringe Filter w/ Prefilter, Hydrophilic PTFE, 13 mm dia., 0.45 µm; 100/pk
Call For Price
COMPARE
0.22 Syringe Filter w/ Prefilter, Hydrophilic PTFE, 25 mm dia., 0.22 µm; 100/pk
Call For Price
COMPARE
0.45 Syringe Filter w/ Prefilter, Hydrophilic PTFE, 25 mm dia., 0.45 µm; 100/pk
Call For Price
COMPARE
0.22 Syringe Filter w/ Prefilter, Hydrophilic PTFE, 30 mm dia., 0.22 µm; 100/pk
Call For Price
COMPARE
0.45 Syringe Filter w/ Prefilter, Hydrophilic PTFE, 30 mm dia., 0.45 µm; 100/pk
Call For Price
COMPARE
0.22 Syringe Filter w/ Prefilter, Hydrophilic PTFE, 25 mm dia., 0.22 µm; 500/pk
Call For Price
COMPARE
0.45 Syringe Filter w/ Prefilter, Hydrophilic PTFE, 25 mm dia., 0.45 µm; 500/pk
Call For Price
COMPARE
0.22 Syringe Filter w/ Prefilter, Hydrophilic PTFE, 30 mm dia., 0.22 µm; 500/pk
Call For Price
COMPARE
0.45 Syringe Filter w/ Prefilter, Hydrophilic PTFE, 30 mm dia., 0.45 µm; 500/pk
Call For Price
COMPARE
0.22 Syringe Filter w/ Prefilter, Hydrophilic PTFE, 13 mm dia., 0.22 µm; 1000/pk
Call For Price
COMPARE
0.45 Syringe Filter w/ Prefilter, Hydrophilic PTFE, 13 mm dia., 0.45 µm; 1000/pk
Call For Price
Showing 1- 12 of 12