Kinesis KX Nonsterile Syringe Filters, Glass Fiber

Borosilicate glass fiber membranes eliminate particulates interfering with UV/VIS spectrophotometric analysis
  • Great for high output pre-filtration of particulate-laden and viscous samples
  • Solvent resistant filter housing with nominal extractable
  • Leak-free Luer-lok® and Luer-Slip connections
  • Color-coded filters for easy organization
Prepare reliable aqueous, organic, and biomolecular samples with membrane filtration inside a polypropylene (PP) housing. Each housing comes with a color-coded retainer ring seal to prevent leaks and sample loss, and the color helps to visually show you the membrane type inside the housing.

Syringe filters have Luer-lok® and Luer-Slip connections for easy, quick and tight connections to tubing, syringes, or needles. Filter packs are available in a resealable storage container to prevent contamination during multiple opening and closings.
$118.25 - $844.00USD / Pkg of 100

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pore Size (µm) Membrane Diameter (mm) Description
Availability Pricing
COMPARE
1 13 Syringe Filter, Glass Fiber, 13 mm dia., 1 µm; 100/pk
$118.25USD / Pkg of 100
COMPARE
1 13 Syringe Filter, Glass Fiber, 13 mm dia., 1 µm; 1000/pk
$844.00USD / Pkg of 1000
COMPARE
1 25 Syringe Filter, Glass Fiber, 25 mm dia., 1 µm; 100/pk
$143.00USD / Pkg of 100
COMPARE
1 25 Syringe Filter, Glass Fiber, 25 mm dia., 1 µm; 500/pk
$545.00USD / Pkg of 500
COMPARE
1 30 Syringe Filter, Glass Fiber, 30 mm dia., 1 µm; 100/pk
$127.25USD / Pkg of 100
COMPARE
1 30 Syringe Filter, Glass Fiber, 30 mm dia., 1 µm; 500/pk
$528.50USD / Pkg of 500
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat