Kinesis KX Soxhlet Thimbles
Easily separate soluble substances from a mixture of solids by extraction
  • Manufactured from pure cellulose, neutral cotton
  • Fat and impurity free
  • Ideal for environmental and food industries
Use these soxhlet thimbles in applications where the original sample is solid in nature—ideal for extraction of fats or pesticides in food and soil materials. One-piece molded design provides excellent mechanical strength and rigidity, ensuring retention of the sample in the thimble throughout the extraction process.
$59.00 - $650.00USD / Pkg of 25
25 variations of this product are available.
See All Product Options Below
25 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (mm) Internal Diameter (mm)
Availability Pricing
COMPARE
80 25
$59.00USD / Pkg of 25
COMPARE
60 30
$67.50USD / Pkg of 25
COMPARE
165 46
$126.00USD / Pkg of 25
COMPARE
37 18
$59.00USD / Pkg of 25
COMPARE
80 22
$59.00USD / Pkg of 25
COMPARE
60 26
$64.00USD / Pkg of 25
COMPARE
170 54
$249.00USD / Pkg of 25
COMPARE
200 70
$326.00USD / Pkg of 25
COMPARE
80 30
$69.00USD / Pkg of 25
COMPARE
130 33
$69.00USD / Pkg of 25
COMPARE
123 41
$84.00USD / Pkg of 25
COMPARE
170 58
$249.00USD / Pkg of 25
COMPARE
250 80
$650.00USD / Pkg of 25
COMPARE
240 64
$283.00USD / Pkg of 25
COMPARE
80 27
$67.50USD / Pkg of 25
COMPARE
100 30
$69.00USD / Pkg of 25
COMPARE
130 37
$81.00USD / Pkg of 25
COMPARE
150 41
$84.00USD / Pkg of 25
COMPARE
130 46
$126.00USD / Pkg of 25
COMPARE
80 26
$64.00USD / Pkg of 25
Showing 1 - 20 of 25