This product is now discontinued, View Related Products
Kinesis KX VaporSafe™ HPLC Solvent Waste Collection Systems
One system to protect your staff and environment from harmful solvent vapor waste
  • One system to maintain pure and clean HPLC mobile phases
  • Leak-proof your waste lines and ports with gas-tight seal connections
  • No more spills from open waste solvent containers in the workplace
  • Stop leaks from mismatched waste line sleeved tubing
  • Seal harmful vapors in a carbon activated filter with a visual full-level indicator
  • Quick-disconnect manifold to rapidly make changes to the HPLC system
Don’t compromise your analytical results with contamination or solvent evaporation. This system protects your findings as it preserves a clean environment! Create a reliable closed HPLC waste container system with no possible leaks. Each system securely connects to your workplace’s HPLC connection ports.
Waste manifold is uniquely designed to completely seal vapors from the environment. All the harmful waste accumulates inside a compact activated carbon filter. A color indicator alerts you when to replace the filter. The manifold offers quick-disconnect connections for easy solvent changes as well as to prevent tangled lines.
Fittings included with kit: (3) Chromatography 1/8" OD plug, (1) Chromatography 2.9 mm OD plug, (2) Chromatography 2.2 mm OD plug, (2) Chromatography 1/16" OD plug, (6) 1/4-28 plugs, (1) 1/2" NPT plug, (1) Filter adapter 1/2" to 1/4" NPT, (1) Organic carbon activated filter with indicator, (3) 1/4" NPT plug, (3) Hose barb adapter 1/4" ID tube, 1/4" NPT, (1) Hose barb adapter 3/8" ID tube, 1/4" NPT, (1) Hose barb adapter 3/16" ID tube, 1/4" NPT, (1) Hose barb adapter 1/8" ID tube, 1/4" NPT, (1) Hose barb Y adapter 1/4" ID, (1) 5"L PTFE tubing with compression union 1/8" OD and 1/8" OD chromatography
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (L)
Availability Pricing
COMPARE
5
Call For Price
COMPARE
10
Call For Price
COMPARE
20