Cole-Parmer® TELOS® Alumina SPE Columns
Easily isolate compounds from nonpolar solvents
  • Polar SPE sorbents
Alumina (acidic, basic, and neutral) are polar SPE sorbents commonly used to isolate compounds from nonpolar solvents. Columns are made of polypropylene and feature Luer-slip outlet.
$135.00 - $200.00USD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sorbent Total Volume Capacity (mL) Bed Mass (mg)
Availability Pricing
COMPARE
Alumina—acidic (AL-A) 1 50
$200.00USD / Pkg of 100
COMPARE
Alumina—neutral (AL-N) 6 1000
$135.00USD / Pkg of 30